siRNA Details
Gene Symbol: 4930520A20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015849.1 | - | 1 | 501 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715395.1 | - | 1 | 499 | Chr. 18 - 82386869 - 82387583 [-] on Build GRCm38 |
