siRNA Details
Gene Symbol: 4931419H13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040593.1 | 4 | 432 | |||
GenBank mRNA | AK016464.1 | - | 4 | 432 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716138.1 | - | 4 | 425 | Chr. 3 - 55055246 - 55083975 [+] on Build GRCm38 |
