siRNA Details
Gene Symbol: 4931418L13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016462.1 | - | 1 | 486 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716135.1 | - | 1 | 486 | Chr. 11 - 101347973 - 101348573 [+] on Build GRCm38 |
