siRNA Details
Gene Symbol: 4931420L22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040561.1 | 3 | 307 | |||
GenBank mRNA | AK016468.1 | - | 3 | 308 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4931420L22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040561.1 | 3 | 307 | |||
GenBank mRNA | AK016468.1 | - | 3 | 308 |