siRNA Details
Gene Symbol: Gm34424 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042051.1 | - | 1 | 71 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB638832.1 | - | 1 | 71 | Chr. 9 - 61509723 - 61519715 [+] on Build GRCm38 |
