siRNA Details
Gene Symbol: Gm34424 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042051.1 | - | 5 | 1403 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB235542.2 | - | 1 | 316 | Chr. 9 - 61533682 - 61534249 [+] on Build GRCm38 |
