siRNA Details
Gene Symbol: Gm30094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC053977.1 | - | 3, 4 | 465 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE632065.1 | - | 3, 4 | 453 | Chr. 18 - 65863548 - 65894755 [-] on Build GRCm38 |
