siRNA Details
Gene Symbol: 1700015P03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006004.1 | - | 1 | 49 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706130.1 | - | 1 | 46 | Chr. 16 - 78449435 - 78450121 [-] on Build GRCm38 |
