siRNA Details
Gene Symbol: AW549542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045702.1 | 2 | 238 | |||
GenBank mRNA | AK035112.1 | - | 260 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB627380.1 | - | 3 | 260 | Chr. 5 - 119575154 - 119627305 [-] on Build GRCm38 |
