siRNA Details
Gene Symbol: AW549542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045702.1 | 5 | 1742 | |||
GenBank mRNA | AK035112.1 | - | 1764 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB103940.2 | - | 4 | 650 | Chr. 5 - 119570133 - 119574667 [-] on Build GRCm38 |
