siRNA Details
Gene Symbol: E230032D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037811.1 | - | 1 | 31 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB631078.1 | - | 1 | 31 | Chr. 7 - 143628024 - 143628659 [+] on Build GRCm38 |
