siRNA Details
Gene Symbol: Gm13920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039323.1 | - | 1 | 129 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB633096.1 | - | 1 | 129 | Chr. 2 - 102057587 - 102060414 [+] on Build GRCm38 |
