siRNA Details
Gene Symbol: Znrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078666.1 | - | 1737 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB324362.2 | - | 1 | 580 | Chr. 6 - 54890312 - 54890950 [+] on Build GRCm38 |
