siRNA Details
Gene Symbol: C030026O17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021105.1 | - | 1 | 645 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728220.1 | - | 1 | 641 | Chr. 2 - 106732996 - 106733666 [+] on Build GRCm38 |
