siRNA Details
Gene Symbol: C030024C20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021101.1 | - | 1 | 509 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728207.1 | - | 1 | 509 | Chr. 16 - 38533104 - 38533651 [-] on Build GRCm38 |
