siRNA Details
Gene Symbol: 9430087B13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020508.1 | - | 1 | 370 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721897.1 | - | 1 | 370 | Chr. 5 - 127309017 - 127309739 [+] on Build GRCm38 |
