siRNA Details
Gene Symbol: A630023A22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041587.1 | - | 1 | 61 | ||
AW989347.1 | - | 1 | 122 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB638275.1 | - | 1 | 61 | Chr. 14 - 34052633 - 34102693 [-] on Build GRCm38 |
