siRNA Details
Gene Symbol: 4930563I02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC048633.1 | - | 1 | 56 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA463135.1 | - | 1 | 59 | Chr. 14 - 60087071 - 60096089 [+] on Build GRCm38 |
