siRNA Details
Gene Symbol: Gm20744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045744.1 | 4 | 407 | |||
GenBank mRNA | AK085155.1 | - | 4 | 408 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB660423.1 | - | 4 | 408 | Chr. 7 - 82070387 - 82079767 [+] on Build GRCm38 |
