siRNA Details
Gene Symbol: Gm20744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045744.1 | 4 | 2091 | |||
GenBank mRNA | AK085155.1 | - | 4 | 2092 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm20744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045744.1 | 4 | 2091 | |||
GenBank mRNA | AK085155.1 | - | 4 | 2092 |