siRNA Details
Gene Symbol: 5730409L17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086162.1 | - | 1 | 2695 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB522699.2 | - | 1 | 672 | Chr. 6 - 8725128 - 8725816 [-] on Build GRCm38 |
