siRNA Details
Gene Symbol: Atxn7l3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037008.1 | - | 1782 | |||
AK051537.1 | - | 2810 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB458124.2 | - | 1 | 427 | Chr. 10 - 112923872 - 112924525 [-] on Build GRCm38 |
