siRNA Details
Gene Symbol: 4933415b22rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016815.1 | - | 1 | 362 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA315563.1 | - | 1 | 113 | Chr. 5 - 112367461 - 112368242 [+] on Build GRCm38 |
