siRNA Details
Gene Symbol: 4930419G24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040595.1 | 1 | 97 | |||
GenBank mRNA | AK015168.1 | - | 1 | 98 | ||
AK029638.1 | - | 1 | 98 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714714.1 | - | 1 | 98 | Chr. 3 - 33024777 - 33088488 [+] on Build GRCm38 |
