siRNA Details
Gene Symbol: 4833411C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102285.1 | 2 | 442 | |||
GenBank mRNA | AK014677.1 | - | 2 | 353 | ||
AK050245.1 | - | 2 | 443 | |||
BC052524.1 | - | 436 | ||||
BY714173.1 | - | 2 | 353 |
