siRNA Details
Gene Symbol: Gm41829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089320.1 | - | 1 | 1778 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY764273.1 | - | 1 | 264 | Chr. 19 - 29239448 - 29240146 [-] on Build GRCm38 |
