siRNA Details
Gene Symbol: 4930406M16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015108.1 | - | 1 | 86 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714643.1 | - | 1 | 83 | Chr. 17 - 65783773 - 65784266 [+] on Build GRCm38 |
