siRNA Details
Gene Symbol: 1810019N24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007554.1 | - | 2 | 356 | ||
AK019651.1 | - | 1 | 358 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715158.1 | - | 1 | 358 | Chr. 7 - 12949690 - 12950066 [-] on Build GRCm38 |
