siRNA Details
Gene Symbol: 4930405N21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015101.1 | - | 1 | 690 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714634.1 | - | 1 | 688 | Chr. 5 - 113320825 - 113321704 [-] on Build GRCm38 |
