siRNA Details
Gene Symbol: 4930406D18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040543.1 | 3 | 522 | |||
GenBank mRNA | AK015105.1 | - | 3 | 489 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930406D18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040543.1 | 3 | 522 | |||
GenBank mRNA | AK015105.1 | - | 3 | 489 |