siRNA Details
Gene Symbol: 4930553I21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016106.1 | - | 1 | 432 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715710.1 | - | 1 | 427 | Chr. 10 - 8860864 - 8861439 [-] on Build GRCm38 |
