siRNA Details
Gene Symbol: 4930553I04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016105.1 | - | 1 | 526 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715709.1 | - | 1 | 521 | Chr. 5 - 128735360 - 128736025 [-] on Build GRCm38 |
