siRNA Details
Gene Symbol: 1810019D21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040344.1 | 5 | 786 | |||
NR_040345.1 | 4 | 710 | ||||
NR_040346.1 | 4 | 517 | ||||
GenBank mRNA | AK007545.1 | - | 2 | 318 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1810019D21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040344.1 | 5 | 786 | |||
NR_040345.1 | 4 | 710 | ||||
NR_040346.1 | 4 | 517 | ||||
GenBank mRNA | AK007545.1 | - | 2 | 318 |