siRNA Details
Gene Symbol: 1810019D21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040344.1 | 5 | 1044 | |||
NR_040345.1 | 4 | 968 | ||||
NR_040346.1 | 4 | 775 | ||||
GenBank mRNA | AK007545.1 | - | 2 | 576 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF583662.1 | - | 2 | 456 | Chr. 8 - 106137189 - 106138465 [+] on Build GRCm38 |
