siRNA Details
Gene Symbol: Gm41043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029426.1 | - | 1, 2 | 193 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614247.1 | - | 1, 2 | 193 | Chr. 13 - 101544396 - 101548152 [-] on Build GRCm38 |
