siRNA Details
Gene Symbol: Gm8149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085632.1 | - | 107 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB661633.1 | - | 2 | 107 | Chr. 7 - 98895930 - 98898820 [+] on Build GRCm38 |
