siRNA Details
Gene Symbol: 5330423I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019905.1 | - | 1 | 1808 | ||
BB026256.1 | - | 1 | 215 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB024703.1 | - | 1 | 195 | Chr. 9 - 34752942 - 34753247 [+] on Build GRCm38 |
