siRNA Details
Gene Symbol: Gm35630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK140699.1 | - | 435 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB647507.1 | - | 1 | 432 | Chr. 2 - 25634837 - 25635288 [+] on Build GRCm38 |
