siRNA Details
Gene Symbol: 6330563C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018239.1 | - | 1 | 1104 | ||
AK047772.1 | - | 1 | 1066 | |||
BI730484.1 | - | 1 | 351 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 6330563C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018239.1 | - | 1 | 1104 | ||
AK047772.1 | - | 1 | 1066 | |||
BI730484.1 | - | 1 | 351 |