siRNA Details
Gene Symbol: 9430019J16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040635.1 | 2 | 177 | |||
GenBank mRNA | AK020426.1 | - | 2 | 178 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721614.1 | - | 2 | 174 | Chr. 2 - 75436329 - 75444176 [-] on Build GRCm38 |
