siRNA Details
Gene Symbol: 1700074I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006948.1 | - | 1 | 261 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706978.1 | - | 1 | 261 | Chr. 15 - 36164892 - 36165444 [+] on Build GRCm38 |
