siRNA Details
Gene Symbol: 1700074A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006946.1 | - | 1 | 14 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706970.1 | - | 1 | 14 | Chr. 1 - 37299352 - 37299697 [-] on Build GRCm38 |
