siRNA Details
Gene Symbol: 1700051K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006761.1 | - | 1 | 646 | ||
AV266842.1 | - | 1 | 204 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV046107.2 | - | 1 | 254 | Chr. 6 - 134245674 - 134245973 [-] on Build GRCm38 |
