siRNA Details
Gene Symbol: 1700072O05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045733.1 | 1, 2 | 173 | |||
GenBank mRNA | AK006942.1 | - | 1, 2 | 174 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706958.1 | - | 1, 2 | 174 | Chr. 6 - 120554796 - 120574204 [+] on Build GRCm38 |
