siRNA Details
Gene Symbol: Gm16136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005410.1 | - | 1 | 82 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705616.1 | - | 1 | 82 | Chr. 15 - 37232143 - 37233141 [-] on Build GRCm38 |
