siRNA Details
Gene Symbol: Zfp952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050612.1 | - | 2 | 244 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB654278.1 | - | 2 | 240 | Chr. 17 - 32993132 - 32996970 [+] on Build GRCm38 |
