siRNA Details
Gene Symbol: 2900042G08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019333.1 | - | 1 | 442 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712609.1 | - | 1 | 438 | Chr. 13 - 6530599 - 6531243 [+] on Build GRCm38 |
