siRNA Details
Gene Symbol: Gm29684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081638.1 | - | 282 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB649153.1 | - | 4 | 282 | Chr. 10 - 95122160 - 95145825 [-] on Build GRCm38 |
