siRNA Details
Gene Symbol: 1700121I08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007225.1 | - | 1 | 206 | ||
AV209809.1 | - | 1 | 178 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707318.1 | - | 1 | 206 | Chr. 9 - 48495979 - 48496357 [-] on Build GRCm38 |
