siRNA Details
Gene Symbol: BB557941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040356.1 | 2 | 161 | |||
GenBank mRNA | AK163349.1 | - | 2 | 162 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY725773.1 | - | 2 | 162 | Chr. 2 - 57127920 - 57181754 [-] on Build GRCm38 |
